(Pentafluorophenyl)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22430-03
B22430-06
CAAAB22430-03EA
66.99
CAD
CAAAB22430-03
CAAAB22430-06
(Pentafluorophenyl)acetic acid 98+%
(Pentafluorophenyl)acetic acid
Purity: 98+%
CAS No.: 653-21-4
Molecular Formula: C8H3F5O2
Formula Weight: 226.10
Melting Point: 107-110°
Beilstein Reference: 2056612
Formula:
C₈H₃F₅O₂ Boiling Pt: 247 °C (1013 hPa) Melting Pt: 107…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004316 CAS Number: 653-21-4 EINECS: 211-497-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...