4,4'-Diaminobenzanilide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-N-(4-aminophenyl)benzamide

B22419-09 B22419-18
CAAAB22419-09EA 46.93 CAD
CAAAB22419-09 CAAAB22419-18 CAAAB22419-30
4,4'-Diaminobenzanilide 98%
4,4'-Diaminobenzanilide

Purity: 98% CAS No.: 785-30-8 Molecular Formula: C13H13N3O Formula Weight: 227.27 Melting Point: 200-202° Beilstein Reference: 2215469

Formula: C₁₃H₁₃N₃O
MW: 227.27 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00025361
CAS Number: 785-30-8
EINECS: 212-321-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR