2-Methyl-6-nitrobenzoxazole 98%

Supplier: Thermo Scientific Chemicals
Warning

B22415-06 B22415-14
CAAAB22415-06EA 133.67 CAD
CAAAB22415-06 CAAAB22415-14 AAB22415-03
2-Methyl-6-nitrobenzoxazole 98%
2-Methyl-6-nitrobenzoxazole

Purity: 98% CAS No.: 5683-43-2 Molecular Formula: C9H8NO Formula Weight: 146.17 Melting Point: 154-156° MDL No.: MFCD00022848

Formula: C₈H₆N₂O₃
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00022848
CAS Number: 5683-43-2
EINECS: 227-149-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR