2-(Diphenylphosphino)benzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B22389-03 B22389-06 B22389-14
CAAAB22389-03EA 108.23 CAD
CAAAB22389-03 CAAAB22389-06 CAAAB22389-14
2-(Diphenylphosphino)benzoic acid 97%
2-(Diphenylphosphino)benzoic acid

Purity: 97% CAS No.: 17261-28-8 Molecular Formula: C19H15O2P Formula Weight: 306.31 Melting Point: 174-181° MDL No.: MFCD00674024

Formula: C₁₉H₁₅O₂P
Melting Pt: 174…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00674024
CAS Number: 17261-28-8
EINECS: 241-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR