3,5-Dichlorobenzhydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22383-03
B22383-06
CAAAB22383-03EA
51.51
CAD
CAAAB22383-03
CAAAB22383-06
3,5-Dichlorobenzhydrazide 97%
3,5-Dichlorobenzhydrazide
Purity: 97%
CAS No.: 62899-78-9
Molecular Formula: C7H6Cl2N2O
Formula Weight: 205.04
Melting Point: 205-207°
MDL No.: MFCD00833411
Formula:
C₇H₆Cl₂N₂O Melting Pt: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00833411 CAS Number: 62899-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...