4-(Trifluoromethyl)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

B22374-03 B22374-06 B22374-14
CAAAB22374-03EA 118.5 CAD
CAAAB22374-03 CAAAB22374-06 CAAAB22374-14
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid

Purity: 98% CAS No.: 128796-39-4 Molecular Formula: C7H6BF3O2 Formula Weight: 189.93 Melting Point: 238-242° Beilstein Reference: 3544189

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 238…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR