trans-4-Aminocyclohexanol 98+%

Supplier: Thermo Scientific Chemicals
Danger

B22365-06 B22365-14 B22365-22
CAAAB22365-06EA 74.89 CAD
CAAAB22365-06 CAAAB22365-14 CAAAB22365-22
trans-4-Aminocyclohexanol 98+%
trans-4-Aminocyclohexanol

Purity: 98+% CAS No.: 27489-62-9 Molecular Formula: C6H13NO Formula Weight: 115.18 Melting Point: 110-113° UN No.: 3259 MDL No.: MFCD00067698

Formula: C₆H₁₃NO
Boiling Pt: 127 °C (14 -rr)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00067698
CAS Number: 27489-62-9
EINECS: 248-492-8
UN: 3259
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR