4-Phenoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals

B22329-03 B22329-06 B22329-14
CAAAB22329-03EA 51.82 CAD
CAAAB22329-03 CAAAB22329-06 CAAAB22329-14
4-Phenoxybenzaldehyde 98%
4-Phenoxybenzaldehyde

Purity: 98% CAS No.: 67-36-7 Molecular Formula: C13H10O2 Formula Weight: 198.22 Flash Point: 110°(230°F) Refractive Index: 1.6110 Melting Point: 24-25° Boiling Point: 184-186°/14mm Beilstein Reference: 1947841 MDL No.: MFCD00003383

Formula: C₁₃H₁₀O₂
Boiling Pt: 184…186 °C/14 mm
Melting Pt: 24…25 °C
Density: 1.132
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003383
CAS Number: 67-36-7
EINECS: 200-650-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR