Dihexyl sulfide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-n-hexylsulfide, Di-n-hexylsulphide

B22325-14 B22325-22
CAAAB22325-14EA 86.27 CAD
CAAAB22325-14 CAAAB22325-22
Dihexyl sulfide 97%
Dihexyl sulphide

Purity: 97% CAS No.: 6294-31-1 Molecular Formula: [CH3(CH2)5]2S Formula Weight: 202.40 Flash Point: 116°(241°F) Refractive Index: 1.4587 Odor: Stench Boiling Point: 260° Beilstein Reference: 1738333 UN No.: 3334 MDL No.: MFCD00009528

Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 260 °C
Melting Pt: –28.3 °C
Density: 0.849
Flash Pt: 116 °C(241 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009528
CAS Number: 6294-31-1
EINECS: 228-556-1
UN: 2810
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR