trans-2-Aminocyclohexanol hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22324-06
B22324-14
B22324-22
CAAAB22324-06EA
139.04
CAD
CAAAB22324-06
CAAAB22324-14
CAAAB22324-22
trans-2-Aminocyclohexanol hydrochloride 98+%
trans-2-Aminocyclohexanol hydrochloride
Purity: 98+%
CAS No.: 5456-63-3
Molecular Formula: C6H13NO.HCl
Formula Weight: 151.64
Melting Point: 172-175°
MDL No.: MFCD00001490
Formula:
C₆H₁₄ClNO Melting Pt: 172…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001490 CAS Number: 5456-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...