ω-Bromo-2,4-dimethoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-2',4'-dimethoxyacetophenone, 2,4-Dimethoxyphenacyl bromide, 2-Bromo-1-(2,4-dimethoxyphenyl)ethan-1-one, 2-Bromo-1-(2,4-dimethoxyphenyl)ethanone, α-Bromo-2,4-dimethoxyacetophenone
B22300-03
B22300-06
CAAAB22300-03EA
143.78
CAD
CAAAB22300-03
CAAAB22300-06
CAAAB22300-14
ω-Bromo-2,4-dimethoxyacetophenone 98%
ω-Bromo-2,4-dimethoxyacetophenone
Purity: 98%
CAS No.: 60965-26-6
Molecular Formula: C10H11BrO3
Formula Weight: 259.10
Storage Temperature: 2°C to 8°C
Melting Point: 102-105°
UN No.: 3261
MDL No.: MFCD00000197
Formula:
C₁₀H₁₁BrO₃ Boiling Pt: 315 °C (1013 hPa) Melting Pt: 102…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000197 CAS Number: 60965-26-6 EINECS: 262-542-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...