4-Propoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

B22266-14 B22266-22
CAAAB22266-06EA 154.4 CAD
CAAAB22266-06 CAAAB22266-14 CAAAB22266-22
4-Propoxyphenol 98%
4-Propoxyphenol

Purity: 98% CAS No.: 18979-50-5 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 110°(230°F) Melting Point: 55-58° MDL No.: MFCD00002335

Formula: C₉H₁₂O₂
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 55…58 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002335
CAS Number: 18979-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR