4,4'-Biphenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22240-14
B22240-22
B22240-36
CAAAB22240-14EA
140.62
CAD
CAAAB22240-14
CAAAB22240-22
CAAAB22240-36
4,4'-Biphenol 99%
4,4'-Biphenol
Purity: 99%
CAS No.: 92-88-6
Molecular Formula: C12H10O2
Formula Weight: 186.21
Odor: Odorless
Melting Point: 279-285°
Beilstein Reference: 1908886
MDL No.: MFCD00002348
Formula:
C₁₂H₁₀O₂ Boiling Pt: 355 °C (1013 hPa) Melting Pt: 279…285 °C Density: 1.25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002348 CAS Number: 92-88-6 EINECS: 202-200-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...