Fuchsin (acid)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzenesulfonic acid, 2-amino-5-[(4-amino-3-sulfophenyl)(4-imino-3-sulfo-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2), Rubin S, CN DcR-8, Acid Violet 19, 3-(1-(4-Amino-3-methyl-5-sulfonatophenyl)-1-(4-amino-3-sulfonatophenyl)methylene)cyclohexa-1,4-dienesulfonic acid, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
CAS No.: 3244-88-0
Molecular Formula: C20H17N3Na2O9S3
Formula Weight: 585.53
Beilstein Reference: 4111656
Merck Reference: 14,107
MDL No.: MFCD00013286
Formula:
C₂₀H₁₇N₃Na₂O₉S₃ Melting Pt: > 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013286 CAS Number: 3244-88-0 EINECS: 221-816-5 Merck Index: 14,107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...