4-Fluoro-2-methoxyphenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22211-06
B22211-14
CAAAAB22211-03EA
94.71
CAD
CAAAAB22211-03
CAAAB22211-06
CAAAB22211-14
4-Fluoro-2-methoxyphenol 97%
4-Fluoro-2-methoxyphenol
Purity: 97%
CAS No.: 450-93-1
Molecular Formula: C7H7FO2
Formula Weight: 142.13
Flash Point: 102°(216°F)
Refractive Index: 1.5170
Odor: Phenol-like
Boiling Point: 195°
Beilstein Reference: 5425200
MDL No.: MFCD00070797
Formula:
C₇H₇FO₂ MW: 142.13 g/mol Boiling Pt: 195 °C Density: 1.25 Flash Pt: 102 °C(216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070797 CAS Number: 450-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...