3-Chloro-2-methylanisole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22203-06
B22203-14
B22203-22
CAAAB22203-06EA
74.1
CAD
CAAAB22203-06
CAAAB22203-14
CAAAB22203-22
3-Chloro-2-methylanisole 97%
3-Chloro-2-methylanisole
Purity: 97%
CAS No.: 3260-88-6
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Flash Point: 92°(197°F)
Refractive Index: 1.5395
Melting Point: -3°
Boiling Point: 212-215°
Beilstein Reference: 1862220
UN No.: 1993
MDL No.: MFCD00070772
Formula:
C₈H₉ClO Boiling Pt: 212…215 °C Melting Pt: –3 °C Density: 1.160 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070772 CAS Number: 3260-88-6 EINECS: 221-862-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...