α,α,α',α'-Tetrafluoro-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α,α,β,β-Tetrafluoro-p-xylene, 1,4-Bis(difluoromethyl)benzene

B22193-03 B22193-06
CAAAB22193-03EA 62.41 CAD
CAAAB22193-03 CAAAB22193-06
α,α,α',α'-Tetrafluoro-p-xylene 98%
α,α,α',α'-Tetrafluoro-p-xylene

Purity: 98% CAS No.: 369-54-0 Molecular Formula: C8H6F4 Formula Weight: 178.13 Flash Point: 63°(145°F) Boiling Point: 83-84°/30mm MDL No.: MFCD01320708

Formula: C₈H₆F₄
MW: 178.13 g/mol
Boiling Pt: 83…84 °C/30 mm
Density: 1.312
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD01320708
CAS Number: 369-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR