2-Chloro-4'-fluoroacetophenone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone

B22188-14 B22188-22
CAAAB22188-14EA 62.88 CAD
CAAAB22188-14 CAAAB22188-22 CAAAB22188-36
2-Chloro-4'-fluoroacetophenone 99%
2-Chloro-4'-fluoroacetophenone

Purity: 99% CAS No.: 456-04-2 Molecular Formula: C8H6ClFO Formula Weight: 172.59 Flash Point: 110°(230°F) Melting Point: 46-50° Boiling Point: 247° Beilstein Reference: 637860 UN No.: 3261 MDL No.: MFCD00011652

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 247 °C
Melting Pt: 46…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011652
CAS Number: 456-04-2
EINECS: 207-256-4
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR