4-Bromo-2-chloro-1-iodobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22185-03
B22185-06
CAAAB22185-03EA
113.44
CAD
CAAAB22185-03
CAAAB22185-06
4-Bromo-2-chloro-1-iodobenzene 97%
1-Bromo-3-chloro-4-iodobenzene
Purity: 97%
CAS No.: 31928-47-9
Molecular Formula: C6H3BrClI
Formula Weight: 317.36
Refractive Index: 1.6722
Melting Point: 33-35°
Boiling Point: 103-105°/1mm
Beilstein Reference: 2435075
MDL No.: MFCD00079709
Formula:
C₆H₃BrClI Boiling Pt: 103…105 °C/1 mm Melting Pt: 33…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079709 CAS Number: 31928-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...