4-(Hydroxymethyl)benzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Carboxybenzylalcohol, HMBA Linker, HMBA
B22183-03
B22183-06
B22183-14
CAAAB22183-03EA
100.17
CAD
CAAAB22183-03
CAAAB22183-06
CAAAB22183-14
4-(Hydroxymethyl)benzoic acid 98+%
4-(Hydroxymethyl)benzoic acid
Purity: 98+%
CAS No.: 3006-96-0
Molecular Formula: C8H8O3
Formula Weight: 152.15
Storage Temperature: 2°C to 8°C
Odor: Odorless
Beilstein Reference: 2690023
MDL No.: MFCD00017598
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017598 CAS Number: 3006-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...