Polyethylene glycol 2,000

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000

B22181-0B B22181-30
CAAAB22181-0BEA 233.84 CAD
CAAAB22181-0B CAAAB22181-30
Polyethylene glycol 2,000
Polyethylene glycol 2,000

CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Odor: Odorless pH: 5-7 Melting Point: 53-55° Merck Reference: 14,7568 MDL No.: MFCD01779611

Formula: H(OCH₂CH₂)nOH
MW: 2000 g/mol
Melting Pt: 53…55 °C
Flash Pt: 250
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR