4-Amino-2,6-dimethylpyrimidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22175-09
B22175-88
CAAAB22175-09EA
114.39
CAD
CAAAB22175-09
CAAAB22175-88
4-Amino-2,6-dimethylpyrimidine 99%
4-Amino-2,6-dimethylpyrimidine
Purity: 99%
CAS No.: 461-98-3
Molecular Formula: C6H9N3
Formula Weight: 123.16
Melting Point: 184-186°
Merck Reference: 14,5326
MDL No.: MFCD00006106
Formula:
C₆H₉N₃ MW: 123.16 Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006106 CAS Number: 461-98-3 EINECS: 207-320-1 Merck Index: 14,5326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...