2'-Hydroxy-4'-methoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

B22173-06 B22173-14 B22173-22
CAAAB22173-06EA 83.9 CAD
CAAAB22173-06 CAAAB22173-14 CAAAB22173-22
2'-Hydroxy-4'-methoxyacetophenone 99%
2'-Hydroxy-4'-methoxyacetophenone

Purity: 99% CAS No.: 552-41-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 1282794 MDL No.: MFCD00008730

Formula: C₉H₁₀O₃
Boiling Pt: 154 °C (20 -rr)
Melting Pt: 48…50 °C
Density: 1.1604 g/cm³ (40 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008730
CAS Number: 552-41-0
EINECS: 209-012-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR