(4-Bromophenyl)acetic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate

B22165-06 B22165-14 B22165-22
CAAAB22165-06EA 72.21 CAD
CAAAB22165-06 CAAAB22165-14 CAAAB22165-22
(4-Bromophenyl)acetic acid ethyl ester 98%
(4-Bromophenyl)acetic acid ethyl ester

Purity: 98% CAS No.: 14062-25-0 Molecular Formula: C10H11BrO2 Formula Weight: 243.10 Melting Point: 29-31° Boiling Point: 88-90°/0.35mm MDL No.: MFCD00016333

Formula: C₁₀H₁₁BrO₂
MW: 243.1 g/mol
Boiling Pt: 88…90 °C/0.35 mm
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00016333
CAS Number: 14062-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR