4'-Methoxychalcone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Methoxyphenyl)-3-phenylprop-2-en-1-one, α-Styryl p-anisyl ketone, 4'-Methoxybenzylidenacetophenone

B22158-06 B22158-14 B22158-22
CAAAAB22158-06EA 46.61 CAD
CAAAAB22158-06 CAAAB22158-14 CAAAB22158-22
4'-Methoxychalcone 97%
4'-Methoxychalcone

Purity: 97% CAS No.: 959-23-9 Molecular Formula: C16H14O2 Formula Weight: 238.29 Melting Point: 101-103° Beilstein Reference: 644640

Formula: C₁₆H₁₄O₂
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00008407
CAS Number: 959-23-9
EINECS: 213-495-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR