Polyethylene glycol 1,000

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 1,000, PEG 900-1100

B22134-0B B22134-30
CAAAB22134-0BEA 113.92 CAD
CAAAB22134-0B CAAAB22134-30
Polyethylene glycol 1,000
Polyethylene glycol 1,000

CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Flash Point: 260°(500°F) Physical Form: Powder/low melting solid Density: 1.101 Appearance: White Odor: Odorless pH: 5-7 Melting Point: 35-40° Merck Reference: 14,7568 MDL No.: MFCD01779605

Formula: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Melting Pt: 35…40 °C
Density: 1.101
Flash Pt: 260 °C(500 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR