2-Pentylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22125-06
B22125-14
CAAAB22125-06EA
104.75
CAD
CAAAB22125-06
CAAAB22125-14
2-Pentylpyridine 98%
2-Pentylpyridine
Purity: 98%
CAS No.: 2294-76-0
Molecular Formula: C10H15N
Formula Weight: 149.24
Flash Point: 79°(174°F)
Refractive Index: 1.4890
Odor: Pleasant
Boiling Point: 203-204°
Beilstein Reference: 2772
MDL No.: MFCD00051828
Formula:
C₁₀H₁₅N Boiling Pt: 203…204 °C Density: 0.903 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051828 CAS Number: 2294-76-0 EINECS: 218-937-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...