Pyrazine-2,3-dicarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22108-06
B22108-14
CAAAB22108-06EA
65.73
CAD
CAAAB22108-06
CAAAB22108-14
Pyrazine-2,3-dicarboxylic acid 98%
Pyrazine-2,3-dicarboxylic acid
Purity: 98%
CAS No.: 89-01-0
Molecular Formula: C6H4N2O4
Formula Weight: 168.11
Melting Point: ca 188° dec.
Beilstein Reference: 147982
Merck Reference: 14,7958
MDL No.: MFCD00006131
Formula:
C₆H₄N₂O₄ Melting Pt: ∼ 188 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006131 CAS Number: 89-01-0 EINECS: 201-875-3 Merck Index: 14,7958 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...