(2,4-Dinitrophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B22102-14 B22102-22
CAAAB22102-14EA 123.24 CAD
CAAAB22102-14 CAAAB22102-22
(2,4-Dinitrophenyl)acetic acid 98%
(2,4-Dinitrophenyl)acetic acid

Purity: 98% CAS No.: 643-43-6 Molecular Formula: C8H6N2O6 Formula Weight: 226.15 Melting Point: 177-182° Beilstein Reference: 1990798

Formula: (C₆H₃(NO₂)₂)CH₂COOH
MW: 226.15 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00007227
CAS Number: 643-43-6
EINECS: 211-398-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR