2-Fluoro-5-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22100-06
B22100-14
CAAAB22100-03EA
38.99
CAD
CAAAB22100-03
CAAAB22100-06
CAAAB22100-14
2-Fluoro-5-nitrobenzoic acid 98%
2-Fluoro-5-nitrobenzoic acid
Purity: 98%
CAS No.: 7304-32-7
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 142-144°
Beilstein Reference: 1912835
Formula:
C₇H₄FNO₄ Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134238 CAS Number: 7304-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...