4-Amino-2-chloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22098-14
B22098-06
CAAAAB22098-14EA
312.84
CAD
CAAAAB22098-14
CAAAB22098-03
CAAAB22098-06
4-Amino-2-chloropyridine 98%
4-Amino-2-chloropyridine
Purity: 98%
CAS No.: 14432-12-3
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Odor: Unpleasant
Melting Point: 91-94°
Boiling Point: 153°/5mm
Beilstein Reference: 108671
MDL No.: MFCD00060089
Formula:
C₅H₅ClN₂ Boiling Pt: 153 °C/5 mm Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060089 CAS Number: 14432-12-3 EINECS: 238-403-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...