3'-Bromo-4'-fluoroacetophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

B22091-06 B22091-14
CAAAB22091-06EA 106.49 CAD
CAAAB22091-06 CAAAB22091-14
3'-Bromo-4'-fluoroacetophenone 98+%
3'-Bromo-4'-fluoroacetophenone

Purity: 98+% CAS No.: 1007-15-4 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Flash Point: 110°(230°F) Melting Point: 55-57° Boiling Point: 150°/12mm Beilstein Reference: 3088698 MDL No.: MFCD00042466

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C/12 mm
Melting Pt: 55…57 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042466
CAS Number: 1007-15-4
EINECS: 213-750-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR