Sodium tridecyl sulfate 99% may cont. up to ca 2% water and residual solvent

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tridecyl sulfuric acid salt, Tridecyl sulphuric acid salt

B22089-03 B22089-06 B22089-14
CAAAB22089-03EA 86.9 CAD
CAAAB22089-03 CAAAB22089-06 CAAAB22089-14
Sodium tridecyl sulfate 99% may cont. up to ca 2% water and residual solvent
Sodium tridecyl sulphate

Purity: 99% CAS No.: 3026-63-9 Molecular Formula: C13H27NaO4S Formula Weight: 302.40 Melting Point: 185-189° MDL No.: MFCD00024993

Formula: C₁₃H₂₇NaO₄S
MW: 302.41
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00024993
CAS Number: 3026-63-9
EINECS: 221-188-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR