3'-Nitroacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

B22062-06 B22062-14 B22062-22
CAAAB22062-06EA 53.25 CAD
CAAAB22062-06 CAAAB22062-14 CAAAB22062-22
3'-Nitroacetanilide 98+%
3'-Nitroacetanilide

Purity: 98+% CAS No.: 122-28-1 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 151-153° Beilstein Reference: 2211961 Merck Reference: 14,6581 MDL No.: MFCD00017015

Formula: C₈H₈N₂O₃
Melting Pt: 151…153 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017015
CAS Number: 122-28-1
EINECS: 204-532-6
Merck Index: 14,6581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR