Hexakis(bromomethyl)benzene 98%

Supplier: Thermo Scientific Chemicals
Danger

B22016-03 B22016-06
CAAAB22016-03EA 84.37 CAD
CAAAB22016-03 CAAAB22016-06
Hexakis(bromomethyl)benzene 98%
Hexakis(bromomethyl)benzene

Purity: 98% CAS No.: 3095-73-6 Molecular Formula: C12H12Br6 Formula Weight: 635.65 Melting Point: ca 292° dec. UN No.: 3261 MDL No.: MFCD00182539

Formula: C₁₂H₁₂Br₆
Melting Pt: ∼ 292 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00182539
CAS Number: 3095-73-6
EINECS: 221-443-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR