Dioctadecyl disulfide 98%
Supplier: Thermo Scientific Chemicals
B21978-03
B21978-06
B21978-14
CAAAB21978-03EA
77.26
CAD
CAAAB21978-03
CAAAB21978-06
CAAAB21978-14
Dioctadecyl disulfide 98%
Dioctadecyl disulphide
Purity: 98%
CAS No.: 2500-88-1
Molecular Formula: CH3(CH2)17S-S(CH2)17CH3
Formula Weight: 571.11
Odor: Unpleasant
Melting Point: 60-64°
Beilstein Reference: 1715376
MDL No.: MFCD00026687
Formula:
C₃₆H₇₄S₂ Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026687 CAS Number: 2500-88-1 EINECS: 219-702-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...