3-Mercapto-1-hexanol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Mercaptohexan-1-ol

B21965-06 B21965-14
CAAAB21965-06EA 129.56 CAD
CAAAB21965-06 CAAAB21965-14
3-Mercapto-1-hexanol 96%
3-Mercapto-1-hexanol

Purity: 96% CAS No.: 51755-83-0 Molecular Formula: C6H14OS Formula Weight: 134.24 Flash Point: 100°(212°F) Refractive Index: 1.4800 Boiling Point: 250° Beilstein Reference: 1846873 MDL No.: MFCD00792515

Formula: C₆H₁₄OS
MW: 134.24 g/mol
Boiling Pt: 250 °C
Density: 0.974
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00792515
CAS Number: 51755-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR