2-Octynoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21957-06
B21957-22
CAAAB21957-06EA
149.15
CAD
CAAAB21957-06
CAAAB21957-22
2-Octynoic acid 98%
2-Octynoic acid
Purity: 98%
CAS No.: 5663-96-7
Molecular Formula: CH3(CH2)4C≡CCO2H
Formula Weight: 140.18
Flash Point: 101°(214°F)
Refractive Index: 1.4600
Boiling Point: 148-149°/19mm
UN No.: 3265
MDL No.: MFCD00004364
Formula:
C₈H₁₂O₂ Boiling Pt: 148…149 °C/19 mm Melting Pt: 5 °C Density: 0.961 Flash Pt: 101 °C (214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004364 CAS Number: 5663-96-7 EINECS: 227-121-3 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...