Polyethylene glycol 10,000
Supplier: Thermo Scientific Chemicals
Synonyms:
PEG 10000, Ethylene glycol polymer 10000, PEG 10,000
B21955-0B
B21955-30
CAAAB21955-0BEA
145.83
CAD
CAAAB21955-0B
CAAAB21955-30
Polyethylene glycol 10,000
Polyethylene glycol 10,000
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 265°(509°F)
Physical Form: Powder
Density: 1.2
Odor: Odorless
pH: 5-7
Melting Point: 58-63°
Merck Reference: 14,7568
MDL No.: MFCD00081839
Formula:
H(OCH₂CH₂)nOH Melting Pt: 58…63 °C Density: 1.2 Flash Pt: 265 °C(509 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 203-473-3 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...