4-Iodoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21936-18
B21936-30
CAAAB21936-18EA
117.87
CAD
CAAAB21936-18
CAAAB21936-30
4-Iodoaniline 98%
4-Iodoaniline
Purity: 98%
CAS No.: 540-37-4
Molecular Formula: C6H6IN
Formula Weight: 219.03
Melting Point: 61-64°
Beilstein Reference: 774101
Merck Reference: 14,5027
MDL No.: MFCD00007848
Formula:
C₆H₆IN Boiling Pt: 262 °C (1013 hPa) Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007848 CAS Number: 540-37-4 EINECS: 208-743-4 UN: 2811 ADR: 6.1,III Merck Index: 14,5027 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...