3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS

B21927-03 B21927-06 B21927-14
CAAAB21927-03EA 110.6 CAD
CAAAB21927-03 CAAAB21927-06 CAAAB21927-14
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) 98+%
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteins


Purity: 98+% CAS No.: 75621-03-3 Molecular Formula: C32H58N2O7S Formula Weight: 614.88 Melting Point: 156-158° dec. MDL No.: MFCD00012116

Formula: C₃₂H₅₈N₂O₇S
Melting Pt: 156…158 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012116
CAS Number: 75621-03-3
Merck Index: 13,02052

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR