Trimesic acid trimethyl ester 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Trimethyl 1,3,5-benzenetricarboxylate, Trimethyl benzene-1,3,5-tricarboxylate, 1,3,5-Benzenetricarboxylic acid trimethyl ester
B21926-06
B21926-14
CAAAB21926-06EA
48.82
CAD
CAAAB21926-06
CAAAB21926-14
Trimesic acid trimethyl ester 98%
Trimesic acid trimethyl ester
Purity: 98%
CAS No.: 2672-58-4
Molecular Formula: C12H12O6
Formula Weight: 252.22
Melting Point: 144-147°
Beilstein Reference: 2219698
Formula:
C₁₂H₁₂O₆ MW: 252.22 Melting Pt: 144…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008434 CAS Number: 2672-58-4 EINECS: 220-215-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...