Polyethylene glycol 200
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
PEG 200, Ethylene glycol polymer average Mn 200
B21918-0B
B21918-30
CAAAAB21918-36EA
164.78
CAD
CAAAAB21918-36
CAAAB21918-0B
CAAAB21918-30
Polyethylene glycol 200
Polyethylene glycol 200
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 171°(339°F)
Refractive Index: 1.4590
Odor: Odorless
pH: 5-7
Melting Point: -55 to -40°
Merck Reference: 14,7568
MDL No.: MFCD01779596
Formula:
H(OCH₂CH₂)nOH MW: 200 g/mol Boiling Pt: ? 250 °C (1013 hPa) Melting Pt: –55…–40 °C Density: 1.127 Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01779596 CAS Number: 25322-68-3 EINECS: 203-473-3 Merck Index: 14,7568 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...