2-Aminobenzenesulfonamide 98%

Supplier: Thermo Scientific Chemicals
Warning

B21912-06 B21912-14 B21912-22
CAAAB21912-06EA 132.25 CAD
CAAAB21912-06 CAAAB21912-14 CAAAB21912-22
2-Aminobenzenesulfonamide 98%
2-Aminobenzenesulphonamide

Purity: 98% CAS No.: 3306-62-5 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Odor: Odorless Melting Point: 153-157° MDL No.: MFCD00007932

Formula: C₆H₈N₂O₂S
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00007932
CAS Number: 3306-62-5
EINECS: 221-988-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR