4-Fluoroanthranilic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21907-06
B21907-03
CAAAAB21907-06EA
279.66
CAD
CAAAAB21907-06
CAAAB21907-01
CAAAB21907-03
4-Fluoroanthranilic acid 98%
4-Fluoroanthranilic acid
Purity: 98%
CAS No.: 446-32-2
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Melting Point: 192-196°
Beilstein Reference: 2803665
Formula:
C₇H₆FNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 192…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075553 CAS Number: 446-32-2 EINECS: 207-163-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...