4-Chloro-2-isopropyl-5-methylphenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21906-14
B21906-22
CAAAB21906-14EA
76.47
CAD
CAAAB21906-14
CAAAB21906-22
4-Chloro-2-isopropyl-5-methylphenol 99%
4-Chloro-2-isopropyl-5-methylphenol
Purity: 99%
CAS No.: 89-68-9
Molecular Formula: C10H13ClO
Formula Weight: 184.67
Melting Point: 59-61°
Boiling Point: 259-263°
Merck Reference: 14,2170
MDL No.: MFCD00002326
Formula:
C₁₀H₁₃ClO Boiling Pt: 259…263 °C Melting Pt: 59…61 °C Density: 1.228 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002326 CAS Number: 89-68-9 EINECS: 201-930-1 Merck Index: 14,2170 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...