1-Bromo-2,3-dichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dichlorobromobenzene

B21904-14 B21904-22
CAAAB21904-14EA 57.83 CAD
CAAAB21904-14 CAAAB21904-22
1-Bromo-2,3-dichlorobenzene 98%
1-Bromo-2,3-dichlorobenzene

Purity: 98% CAS No.: 56961-77-4 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Melting Point: 56-60° Boiling Point: 243° MDL No.: MFCD00000536

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 243 °C
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000536
CAS Number: 56961-77-4
EINECS: 260-476-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR