3-Phenoxytoluene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenyl m-tolyl ether, 1-Methyl-3-phenoxybenzene

B21899-14 B21899-22
CAAAB21899-14EA 64.15 CAD
CAAAB21899-14 CAAAB21899-22
3-Phenoxytoluene 97%
3-Phenoxytoluene

Purity: 97% CAS No.: 3586-14-9 Molecular Formula: C13H12O Formula Weight: 184.24 Flash Point: 110°(230°F) Refractive Index: 1.5740 Boiling Point: 271-273° Beilstein Reference: 2045714 MDL No.: MFCD00008531

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 271
Density: 1.052
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008531
CAS Number: 3586-14-9
EINECS: 222-716-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR