6-Methylquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

B21892-06 B21892-14 B21892-22
CAAAAB21892-06EA 41.24 CAD
CAAAAB21892-06 CAAAB21892-14 CAAAB21892-22
6-Methylquinoline 98%
6-Methylquinoline

Purity: 98% CAS No.: 91-62-3 Molecular Formula: C10H9N Formula Weight: 143.19 Flash Point: 110°(230°F) Refractive Index: 1.6150 Melting Point: -22° Boiling Point: 257-259° Beilstein Reference: 110336 MDL No.: MFCD00006804

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 257…259 °C
Melting Pt: –22 °C
Density: 1.066
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006804
CAS Number: 91-62-3
EINECS: 202-084-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR