Florisil® 100-200 mesh for chromatography
Supplier: Thermo Scientific Chemicals
Synonyms:
Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
B21870-0B
B21870-30
CAAAB21870-0BEA
527.72
CAD
CAAAB21870-0B
CAAAB21870-30
Florisil® 100-200 mesh for chromatography
Magnesium silicate
CAS No.: 1343-88-0
MDL No.: MFCD00078249
Formula:
MgO₃Si MW: 100.38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078249 CAS Number: 1343-88-0 EINECS: 215-681-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...